Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzenesulfonyl Chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
98-68-0
C7H7ClO3S
206.65
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride,p-anisylsulfonyl chloride,benzenesulfonyl chloride, 4-methoxy,4-methoxybenzenesulphonyl chloride,4-methoxybenzenesulfonylchloride,4-methoxy-benzenesulfonyl chloride,p-anisolesulfonyl chloride,p-methoxyphenylsulfonyl chloride,4-methoxyphenylsulfonyl chloride,benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(=O)(=O)Cl
98-68-0
C7H7ClO3S
206.65
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride,p-anisylsulfonyl chloride,benzenesulfonyl chloride, 4-methoxy,4-methoxybenzenesulphonyl chloride,4-methoxybenzenesulfonylchloride,4-methoxy-benzenesulfonyl chloride,p-anisolesulfonyl chloride,p-methoxyphenylsulfonyl chloride,4-methoxyphenylsulfonyl chloride,benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(=O)(=O)Cl
Melting Point 40.0°C to 42.0°C
Packaging Glass bottle
Color White to Beige
Quantity 500g
Flash Point >110°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OC6H4SO2Cl
Beilstein 11, 243
Fieser 09,297
Solubility Information Solubility in water: decomposes
Formula Weight 206.65
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material 4-Methoxybenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger