Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzenethiol 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
696-63-9
C7H8OS
140.2
MFCD00004849
NIFAOMSJMGEFTQ-UHFFFAOYSA-N
4-methoxythiophenol,p-methoxybenzenethiol,benzenethiol, 4-methoxy,4-methoxybenzene-1-thiol,benzenethiol, p-methoxy,4-mercaptoanisole,4-methoxy thiophenol,4-methoxy-benzenethiol,para-methoxybenzenethiol,4-mercaptoanisol
12765
4-methoxybenzenethiol
COC1=CC=C(C=C1)S
696-63-9
C7H8OS
140.2
MFCD00004849
NIFAOMSJMGEFTQ-UHFFFAOYSA-N
4-methoxythiophenol,p-methoxybenzenethiol,benzenethiol, 4-methoxy,4-methoxybenzene-1-thiol,benzenethiol, p-methoxy,4-mercaptoanisole,4-methoxy thiophenol,4-methoxy-benzenethiol,para-methoxybenzenethiol,4-mercaptoanisol
12765
4-methoxybenzenethiol
COC1=CC=C(C=C1)S
Specific Gravity 1.14
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5mL
Refractive Index 1.5821 to 1.5841
Flash Point 96°C
Density 1.1400g/mL
Boiling Point 228.0°C
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4SH
Beilstein 06, 859
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethylether,benzene,,chloroform
Formula Weight 140.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzenethiol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 211-799-2

RTECSNumber : DC1800000