Learn More
4-Methoxybenzyl carbazate, 97%, Thermo Scientific Chemicals
Description
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 75°C to 77°C |
Quantity | 5 g |
Beilstein | 2052164 |
Solubility Information | Insoluble in water. |
Formula Weight | 196.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxybenzyl carbazate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 242-664-6 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.