Alfa Aesar™ 4-Methoxybenzyl mercaptan, 98%

Manufacturer: Alfa Aesar™ B2254214

Quantity 25g

 View more versions of this product

Catalog No. AAB2254214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6258-60-2
    Molecular Formula C8H10OS
    Formula Weight 154.23
    MDL Number MFCD00004871
    Beilstein 471686
    Synonym 4-Methoxy-alpha-toluenethiol
    Chemical Name or Material 4-Methoxybenzyl mercaptan

    Specifications

    Formula Weight 154.23
    Molecular Formula C8H10OS
    MDL Number MFCD00004871
    CAS 6258-60-2
    Beilstein 471686
    Synonym 4-Methoxy-alpha-toluenethiol
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Flash Point >110°C (230°F)
    Odor Unpleasant
    Boiling Point 90° to 95°C (0.5mmHg)
    Quantity 25g
    Chemical Name or Material 4-Methoxybenzyl mercaptan
    Assay 98%

    Description

    Applications
    4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Air sensitive. Store under inert gas. Incompatible with oxidizing agents, acids, bases, air.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280h-P305+P351+P338
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit