Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methoxybenzyl)triphenylphosphonium chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
3462-97-3
C26H24ClOP
418.901
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride,4-methoxyphenyl methyl triphenylphosphanium chloride,acmc-1ctv8,4-methoxyphenyl methyl-triphenyl-phosphanium chloride,4-methoxybenzyltriphenylphosphonium chloride,4-methoxybenzyltriphenylphosphonium-chloride,4-methoxybenzyl triphenyphosphonium chloride,p-methoxybenzyl-triphenylphosphonium chloride,4-methoxybenzyltriphenyl phosphonium chloride,4-methoxybenzyl triphenyl phosphonium chloride
2773651
(4-methoxyphenyl)methyl-triphenylphosphanium;chloride
COC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
3462-97-3
C26H24ClOP
418.901
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride,4-methoxyphenyl methyl triphenylphosphanium chloride,acmc-1ctv8,4-methoxyphenyl methyl-triphenyl-phosphanium chloride,4-methoxybenzyltriphenylphosphonium chloride,4-methoxybenzyltriphenylphosphonium-chloride,4-methoxybenzyl triphenyphosphonium chloride,p-methoxybenzyl-triphenylphosphonium chloride,4-methoxybenzyltriphenyl phosphonium chloride,4-methoxybenzyl triphenyl phosphonium chloride
2773651
(4-methoxyphenyl)methyl-triphenylphosphanium;chloride
COC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
For C-C Bond Formation (Olefination)
Packaging Glass Bottle
Melting Point 244.0°C
Quantity 100g
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4CH2P(C6H5)3Cl
Formula Weight 418.9
Physical Form Powder
Percent Purity 98%
Chemical Name or Material (4-Methoxybenzyl)triphenylphosphonium chloride
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.