Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
2393-23-9
C8H11NO
137.18
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine,4-methoxyphenyl methanamine,benzenemethanamine, 4-methoxy,p-methoxybenzylamine,4-methoxy-benzylamine,anisylamine,benzylamine, p-methoxy,4-methoxybenzyl amine,1-4-methoxyphenyl methanamine,4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN
2393-23-9
C8H11NO
137.18
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine,4-methoxyphenyl methanamine,benzenemethanamine, 4-methoxy,p-methoxybenzylamine,4-methoxy-benzylamine,anisylamine,benzylamine, p-methoxy,4-methoxybenzyl amine,1-4-methoxyphenyl methanamine,4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN
Packaging Glass bottle
Refractive Index 1.5460 to 1.5480
Density 1.0500g/mL
Flash Point >110°C
Quantity 500mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 236.0°C to 237.0°C
Specific Gravity 1.05
Assay Percent Range 98%
Linear Formula CH3OC6H4CH2NH2
Beilstein 13,606
Formula Weight 137.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzylamine
Show More Show Less