Alfa Aesar™ 4'-Methoxybiphenyl-4-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5190703

Quantity 1g

 View more versions of this product

Catalog No. AAH5190703

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C19H23BO3
    Formula Weight 310.2
    MDL Number MFCD16294530
    Synonym 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4-(4-Methoxyphenyl)benzeneboronic acid pinacol ester
    Chemical Name or Material 4'-Methoxybiphenyl-4-boronic acid pinacol ester

    Specifications

    Formula Weight 310.2
    Molecular Formula C19H23BO3
    MDL Number MFCD16294530
    Quantity 1g
    Synonym 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4-(4-Methoxyphenyl)benzeneboronic acid pinacol ester
    Chemical Name or Material 4'-Methoxybiphenyl-4-boronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit