Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methoxycarbonyl)phenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass Bottle
99768-12-4
C8H9BO4
179.966
PQCXFUXRTRESBD-UHFFFAOYSA-N
methyl 4-boronobenzoate,4-methoxycarbonyl phenylboronic acid,4-methoxycarbonyl phenyl boronic acid,4-methoxycarbonyl benzeneboronic acid,4-methoxycarbonylphenyl boronic acid,methyl4-boronobenzoate,4-methoxycarbonylbenzeneboronic acid,p-methoxycarbonyl phenylboronic acid,4-methoxycabonyl phenylboronic acid
2734369
(4-methoxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OC)(O)O
99768-12-4
C8H9BO4
179.966
PQCXFUXRTRESBD-UHFFFAOYSA-N
methyl 4-boronobenzoate,4-methoxycarbonyl phenylboronic acid,4-methoxycarbonyl phenyl boronic acid,4-methoxycarbonyl benzeneboronic acid,4-methoxycarbonylphenyl boronic acid,methyl4-boronobenzoate,4-methoxycarbonylbenzeneboronic acid,p-methoxycarbonyl phenylboronic acid,4-methoxycabonyl phenylboronic acid
2734369
(4-methoxycarbonylphenyl)boronic acid
B(C1=CC=C(C=C1)C(=O)OC)(O)O
Packaging Glass Bottle
Quantity 100mg
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 179.97
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-(Methoxycarbonyl)phenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.