Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4′-Methoxycarbonylbenzo-15-crown 5-Ether 96.0+%, TCI America™

Click to view available options
Quantity:
1g
56683-56-8
C16H22O7
326.345
MFCD06797129
PMGUUVHWCHSZKY-UHFFFAOYSA-N
2,3-(4-Methoxycarbonylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
12520240
methyl 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carboxylate
COC(=O)C1=CC2=C(C=C1)OCCOCCOCCOCCO2
56683-56-8
C16H22O7
326.345
MFCD06797129
PMGUUVHWCHSZKY-UHFFFAOYSA-N
2,3-(4-Methoxycarbonylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
12520240
methyl 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carboxylate
COC(=O)C1=CC2=C(C=C1)OCCOCCOCCOCCO2
Color White-Yellow
Melting Point 82°C
Quantity 1g
Formula Weight 326.35
Physical Form Crystal-Powder at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material 4′-Methoxycarbonylbenzo-15-crown 5-Ether