Alfa Aesar™ 4'-Methoxychalcone, 97%

Manufacturer: Alfa Aesar™ B2215822

Quantity 100g

 View more versions of this product

Catalog No. AAB2215822

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 959-23-9
    Molecular Formula C16H14O2
    Formula Weight 238.29
    MDL Number MFCD00008407
    Beilstein 644640
    Synonym 2-Benzylidene-4'-methoxyacetophenone
    Chemical Name or Material 4'-Methoxychalcone

    Specifications

    Formula Weight 238.29
    Molecular Formula C16H14O2
    MDL Number MFCD00008407
    CAS 959-23-9
    Beilstein 644640
    Synonym 2-Benzylidene-4'-methoxyacetophenone
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 101° to 103°C
    Quantity 100g
    Chemical Name or Material 4'-Methoxychalcone
    Assay 97%

    Description

    Applications
    4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

    Solubility
    Soluble in dichloromethane and methanol. Insoluble in water.

    Notes
    Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit