Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxycoumarin, 98%

Click to view available options
Quantity:
5g
25g
100g
20280-81-3
C10H8O3
176.171
MFCD00017349
MLCMXDYMSAZNPC-UHFFFAOYSA-N
4-methoxycoumarin,4-methoxycourmarin,4-methoxy-2h-chromen-2-one,2h-1-benzopyran-2-one, 4-methoxy,2h-1-benzopyran-2-one,4-methoxy,4-methoxy-2h-chromen-2-on,4-methoxycoumarine,acmc-20amex,coumarin, 4-methoxy
529426
4-methoxychromen-2-one
COC1=CC(=O)OC2=CC=CC=C21
20280-81-3
C10H8O3
176.171
MFCD00017349
MLCMXDYMSAZNPC-UHFFFAOYSA-N
4-methoxycoumarin,4-methoxycourmarin,4-methoxy-2h-chromen-2-one,2h-1-benzopyran-2-one, 4-methoxy,2h-1-benzopyran-2-one,4-methoxy,4-methoxy-2h-chromen-2-on,4-methoxycoumarine,acmc-20amex,coumarin, 4-methoxy
529426
4-methoxychromen-2-one
COC1=CC(=O)OC2=CC=CC=C21
Quantity 5g
Melting Point 122°C to 125°C
Beilstein 145960
Formula Weight 176.17
Percent Purity 98%
Chemical Name or Material 4-Methoxycoumarin

TSCA : No

Recommended Storage : Ambient temperatures