Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxymetanilyl Fluoride 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
498-74-8
C7H8FNO3S
205.2
MFCD00007417
SESLQGPHIQXYGF-UHFFFAOYSA-N
4-methoxymetanilyl fluoride,3-amino-4-methoxyphenylsulphonyl fluoride,3-amino-4-methoxy benzene sulphonylfluoride,3-amino-4-methoxyphenyl fluorosulfone,3-amino-4-methoxy-benzenesulfonyl fluoride,3-amino-4-methoxybenzenesulphonyl fluoride,3-azanyl-4-methoxy-benzenesulfonyl fluoride,benzenesulfonylfluoride, 3-amino-4-methoxy,3-amino-4-methoxybenzene-1-sulfonyl fluoride,benzenesulfonyl fluoride, 3-amino-4-methoxy
96952
3-amino-4-methoxybenzenesulfonyl fluoride
COC1=C(C=C(C=C1)S(=O)(=O)F)N
498-74-8
C7H8FNO3S
205.2
MFCD00007417
SESLQGPHIQXYGF-UHFFFAOYSA-N
4-methoxymetanilyl fluoride,3-amino-4-methoxyphenylsulphonyl fluoride,3-amino-4-methoxy benzene sulphonylfluoride,3-amino-4-methoxyphenyl fluorosulfone,3-amino-4-methoxy-benzenesulfonyl fluoride,3-amino-4-methoxybenzenesulphonyl fluoride,3-azanyl-4-methoxy-benzenesulfonyl fluoride,benzenesulfonylfluoride, 3-amino-4-methoxy,3-amino-4-methoxybenzene-1-sulfonyl fluoride,benzenesulfonyl fluoride, 3-amino-4-methoxy
96952
3-amino-4-methoxybenzenesulfonyl fluoride
COC1=C(C=C(C=C1)S(=O)(=O)F)N
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 10g
Melting Point 60°C to 65°C
Color Beige to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H3(NH2)SO2F
Beilstein 14, II, 487
Formula Weight 205.2
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 4-Methoxymetanilyl fluoride, 97%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-870-2