Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxynaphthalene-1-carboxylic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
13041-62-8
C12H10O3
202.209
MFCD02575372
WRQHSQDGYYDRMX-UHFFFAOYSA-N
4-methoxy-1-naphthoic acid,4-methoxy-naphthalene-1-carboxylic acid,1-naphthalenecarboxylic acid, 4-methoxy,1-naphthalenecarboxylicacid, 4-methoxy,4-methoxynaphthalenecarboxylic acid,ksc181m4d,4-methoxy-1-naphthoic acid,4-methoxy-1-naphthoic acid,4-methoxy-1-naphthalenecarboxylic acid
765380
4-methoxynaphthalene-1-carboxylic acid
COC1=CC=C(C2=CC=CC=C21)C(=O)O
13041-62-8
C12H10O3
202.209
MFCD02575372
WRQHSQDGYYDRMX-UHFFFAOYSA-N
4-methoxy-1-naphthoic acid,4-methoxy-naphthalene-1-carboxylic acid,1-naphthalenecarboxylic acid, 4-methoxy,1-naphthalenecarboxylicacid, 4-methoxy,4-methoxynaphthalenecarboxylic acid,ksc181m4d,4-methoxy-1-naphthoic acid,4-methoxy-1-naphthoic acid,4-methoxy-1-naphthalenecarboxylic acid
765380
4-methoxynaphthalene-1-carboxylic acid
COC1=CC=C(C2=CC=CC=C21)C(=O)O
Applications
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Color White
Melting Point 243°C to 247°C
Quantity 25g
Beilstein 2099800
Formula Weight 202.21
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material 4-Methoxynaphthalene-1-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning