Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenethyl alcohol, 96%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
702-23-8
C9H12O2
152.19
MFCD00002900
AUWDOZOUJWEPBA-UHFFFAOYSA-N
2-4-methoxyphenyl ethanol,4-methoxyphenethyl alcohol,4-methoxybenzeneethanol,benzeneethanol, 4-methoxy,p-methoxyphenethyl alcohol,2-4-methoxyphenyl ethan-1-ol,unii-3mwl5nze9k,3mwl5nze9k,4-2-hydroxyethyl anisole,2-4-methoxyphenyl-ethanol
69705
2-(4-methoxyphenyl)ethanol
COC1=CC=C(C=C1)CCO
702-23-8
C9H12O2
152.19
MFCD00002900
AUWDOZOUJWEPBA-UHFFFAOYSA-N
2-4-methoxyphenyl ethanol,4-methoxyphenethyl alcohol,4-methoxybenzeneethanol,benzeneethanol, 4-methoxy,p-methoxyphenethyl alcohol,2-4-methoxyphenyl ethan-1-ol,unii-3mwl5nze9k,3mwl5nze9k,4-2-hydroxyethyl anisole,2-4-methoxyphenyl-ethanol
69705
2-(4-methoxyphenyl)ethanol
COC1=CC=C(C=C1)CCO
Quantity 50g
Boiling Point 334°C to 336°C
Melting Point 28°C to 30°C
Flash Point >112°C
Color Brown
Refractive Index 1.5360 to 1.538
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula CH3OC6H4CH2CH2OH
Beilstein 06, 906
Formula Weight 152.19
Physical Form Liquid After Melting
Percent Purity 96%
Chemical Name or Material 4-Methoxyphenethyl alcohol, 96%
Show More Show Less

EINECSNumber : 211-866-6

TSCA : TSCA