Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride, 97%, Thermo Scientific™

Catalog Number sew03999da
Click to view available options
Quantity:
1 g
25519-82-8
C13H18ClNO2
255.742
MFCD00114817
BBDTWYQCXXFKDH-UHFFFAOYSA-N
4-4-methoxybenzoyl piperidine hydrochloride, 4-methoxyphenyl 4-piperidyl methanone hydrochloride, 4-methoxyphenyl piperidin-4-yl methanone hydrochloride, methanone, 4-methoxyphenyl-4-piperidinyl-, hydrochloride, 4-4-methoxybenzoyl piperidine hcl, 4-4-methoxybenzoyl-piperidine hydrochloride, 4-methoxy-phenyl-piperidin-4-yl-methanone hydrochloride, ksc801q4b, 4-methoxyphenyl-4-piperidyl methanonehcl, 4-methoxyphenyl 4-piperidyl ketone, chloride
2740587
(4-methoxyphenyl)-piperidin-4-ylmethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)C2CCNCC2.Cl
25519-82-8
C13H18ClNO2
255.742
MFCD00114817
BBDTWYQCXXFKDH-UHFFFAOYSA-N
4-4-methoxybenzoyl piperidine hydrochloride, 4-methoxyphenyl 4-piperidyl methanone hydrochloride, 4-methoxyphenyl piperidin-4-yl methanone hydrochloride, methanone, 4-methoxyphenyl-4-piperidinyl-, hydrochloride, 4-4-methoxybenzoyl piperidine hcl, 4-4-methoxybenzoyl-piperidine hydrochloride, 4-methoxy-phenyl-piperidin-4-yl-methanone hydrochloride, ksc801q4b, 4-methoxyphenyl-4-piperidyl methanonehcl, 4-methoxyphenyl 4-piperidyl ketone, chloride
2740587
(4-methoxyphenyl)-piperidin-4-ylmethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)C2CCNCC2.Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 255.74
Percent Purity 97%
Chemical Name or Material (4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit