Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methoxyphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
Specifications
CAS Min % | 97.0 |
Density | 1.1900g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3OC6H4NCS |
Quantity | 10 g |
Formula Weight | 165.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxyphenyl isothiocyanate |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction