Learn More
4-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Description
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
Specifications
Melting Point | 18°C to 20°C |
Density | 1.196 |
Boiling Point | 279°C to 281°C |
Flash Point | >110°C (230°F) |
Odor | Pungent |
Refractive Index | 1.6489 |
Quantity | 50 g |
UN Number | UN2922 |
Beilstein | 606967 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H331-H314-H318-H302-H312 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 218-921-3 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.