Alfa Aesar™ 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 97%

Manufacturer: Alfa Aesar™ H5579206

Quantity 5g

 View more versions of this product

Catalog No. AAH5579206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67589-52-0
    Molecular Formula C19H28O3
    Formula Weight 304.43
    MDL Number MFCD11053432
    Chemical Name or Material 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate

    Specifications

    Formula Weight 304.43
    MDL Number MFCD11053432
    Quantity 5g
    TSCA TSCA
    Molecular Formula C19H28O3
    CAS 67589-52-0
    Melting Point 40° to 44°C
    Chemical Name or Material 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate
    Assay 97%

    Description

    Applications
    4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Sparingly Soluble in water (1.7E-3 g/L) (25°C).

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit