Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylacetamide, 98%, Thermo Scientific™

Click to view available options
Quantity:
5g
6343-93-7
C9H11NO2
165.192
MFCD00017144
OLKQIWCQICCYQS-UHFFFAOYSA-N
2-4-methoxyphenyl acetamide, 4-methoxyphenylacetamide, p-methoxyphenylacetamide, benzeneacetamide, 4-methoxy, 4-methoxyphenoxyphenylacetamide, acmc-20amem, 4-methoxyphe-nylacetamide, benzeneacetamide,4-methoxy, 4-methoxyphenyl-acetamide, para methoxy phenyl acetamide
241868
2-(4-methoxyphenyl)acetamide
COC1=CC=C(C=C1)CC(=O)N
6343-93-7
C9H11NO2
165.192
MFCD00017144
OLKQIWCQICCYQS-UHFFFAOYSA-N
2-4-methoxyphenyl acetamide, 4-methoxyphenylacetamide, p-methoxyphenylacetamide, benzeneacetamide, 4-methoxy, 4-methoxyphenoxyphenylacetamide, acmc-20amem, 4-methoxyphe-nylacetamide, benzeneacetamide,4-methoxy, 4-methoxyphenyl-acetamide, para methoxy phenyl acetamide
241868
2-(4-methoxyphenyl)acetamide
COC1=CC=C(C=C1)CC(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 184°C to 188°C
Beilstein 2088040
Quantity 5g
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenylacetamide

EINECSNumber : 228-745-9

TSCA : No

Recommended Storage : Ambient temperatures