Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylacetyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
4693-91-8
C9H9ClO2
184.62
MFCD00075474
CXJOONIFSVSFAD-UHFFFAOYSA-N
4-methoxyphenylacetyl chloride,2-4-methoxyphenyl acetyl chloride,benzenacetyl chloride, 4-methoxy,benzeneacetyl chloride, 4-methoxy,4-methoxyphenylacetylchloride,p-methoxyphenyl acetyl chloride,p-methoxyphenylacetyl chloride,acmc-1aso1,4-methoxyphenylacetylchlorid,4-methoxybenzeneacetyl chloride
2734688
2-(4-methoxyphenyl)acetyl chloride
COC1=CC=C(C=C1)CC(=O)Cl
4693-91-8
C9H9ClO2
184.62
MFCD00075474
CXJOONIFSVSFAD-UHFFFAOYSA-N
4-methoxyphenylacetyl chloride,2-4-methoxyphenyl acetyl chloride,benzenacetyl chloride, 4-methoxy,benzeneacetyl chloride, 4-methoxy,4-methoxyphenylacetylchloride,p-methoxyphenyl acetyl chloride,p-methoxyphenylacetyl chloride,acmc-1aso1,4-methoxyphenylacetylchlorid,4-methoxybenzeneacetyl chloride
2734688
2-(4-methoxyphenyl)acetyl chloride
COC1=CC=C(C=C1)CC(=O)Cl
Specific Gravity 1.208
Refractive Index 1.5390 to 1.5410
Quantity 2g
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 143.0°C (10.0 mmHg)
Density 1.2080g/mL
Flash Point >110°C
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H4CH2COCl
Beilstein 10, III, 434
Solubility Information Solubility in water: reacts
Formula Weight 184.62
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenylacetyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger