Alfa Aesar™ 4-Methoxyphenylmagnesium bromide, 0.25M in THF

Manufacturer: Alfa Aesar™ H55397AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH55397AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13139-86-1
    Molecular Formula C7H7BrMgO
    Formula Weight 211.34
    MDL Number MFCD00013515
    UN Number UN2924
    Synonym 4-Anisylmagnesium bromide
    Chemical Name or Material 4-Methoxyphenylmagnesium bromide

    Specifications

    Formula Weight 211.34
    Molecular Formula C7H7BrMgO
    MDL Number MFCD00013515
    CAS 13139-86-1
    UN Number UN2924
    Concentration 0.25M in THF
    Synonym 4-Anisylmagnesium bromide
    Quantity 500mL
    Flash Point −20°C (−4°F)
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.
    Chemical Name or Material 4-Methoxyphenylmagnesium bromide

    Description

    Applications
    Used as pharmaceutical intermediate.

    Solubility
    Reacts with water.

    Notes
    Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit