Alfa Aesar™ 4-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF

Manufacturer: Alfa Aesar™ H54317AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54317AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13139-86-1
    Molecular Formula C7H7BrMgO
    Formula Weight 211.34
    MDL Number MFCD00013515
    UN Number UN2924
    Chemical Name or Material 4-Methoxyphenylmagnesium bromide

    Specifications

    Formula Weight 211.34
    Molecular Formula C7H7BrMgO
    MDL Number MFCD00013515
    UN Number UN2924
    Concentration 1.0M in 2-MeTHF
    Quantity 100mL
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13139-86-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material 4-Methoxyphenylmagnesium bromide

    Description

    Applications
    4-Methoxyphenylmagnesium bromide is applied in Grignard-reactions as a reagent for the introduction of the 4-methoxyphenyl group.

    Solubility
    It reacts with water.

    Notes
    Air and moisture sensitive. Incompatible with oxidizing agents and acids.

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit