Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxypropiophenone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
121-97-1
C10H12O2
164.2
MFCD00009310
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
4'-methoxypropiophenone,p-methoxypropiophenone,4-methoxypropiophenone,1-4-methoxyphenyl propan-1-one,1-propanone, 1-4-methoxyphenyl,propiophenone, 4'-methoxy,ethyl 4-methoxyphenyl ketone,1-4-methoxyphenyl-1-propanone,1-propanone,1-4-methoxyphenyl,1-4-methoxy-phenyl-propan-1-one
67144
1-(4-methoxyphenyl)propan-1-one
CCC(=O)C1=CC=C(C=C1)OC
121-97-1
C10H12O2
164.2
MFCD00009310
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
4'-methoxypropiophenone,p-methoxypropiophenone,4-methoxypropiophenone,1-4-methoxyphenyl propan-1-one,1-propanone, 1-4-methoxyphenyl,propiophenone, 4'-methoxy,ethyl 4-methoxyphenyl ketone,1-4-methoxyphenyl-1-propanone,1-propanone,1-4-methoxyphenyl,1-4-methoxy-phenyl-propan-1-one
67144
1-(4-methoxyphenyl)propan-1-one
CCC(=O)C1=CC=C(C=C1)OC
Melting Point 27°C to 29°C
Flash Point 61°C
Boiling Point 273°C to 275°C
Specific Gravity 1.07
Density 1.0700g/mL
Refractive Index 1.5455 to 1.5475
Packaging Glass bottle
Quantity 100g
Color Amber to Colorless
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OC6H4COC2H5
Beilstein 08, 103
Formula Weight 164.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4'-Methoxypropiophenone, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-512-7

TSCA : TSCA