Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxypyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
620-08-6
C6H7NO
109.13
MFCD00674049
XQABVLBGNWBWIV-UHFFFAOYSA-N
pyridine, 4-methoxy,methyl pyridin-4-yl ether,4-methoxy pyridine,.gamma.-methoxypyridine,4-methoxypyridin,4-methoxypyrdine,4-methoxylpyridine,4-methoxy-pyridine,4-methoxypyridine,pubchem2585
69278
4-methoxypyridine
COC1=CC=NC=C1
620-08-6
C6H7NO
109.13
MFCD00674049
XQABVLBGNWBWIV-UHFFFAOYSA-N
pyridine, 4-methoxy,methyl pyridin-4-yl ether,4-methoxy pyridine,.gamma.-methoxypyridine,4-methoxypyridin,4-methoxypyrdine,4-methoxylpyridine,4-methoxy-pyridine,4-methoxypyridine,pubchem2585
69278
4-methoxypyridine
COC1=CC=NC=C1
Refractive Index 1.5180 to 1.5220
Packaging Glass bottle
Density 1.0750g/mL
Color Colorless to Yellow
Boiling Point 191.0°C
Quantity 25mL
Infrared Spectrum Authentic
Melting Point 4.0°C
Flash Point 74°C
Specific Gravity 1.075
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 21,49
Formula Weight 109.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Methoxypyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 210-624-7

Recommended Storage : Darkens on storage