Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methoxystilbene, 98%

Click to view available options
Quantity:
5g
25g
100g
1142-15-0
C15H14O
210.276
MFCD00017178
XWYXLYCDZKRCAD-BQYQJAHWSA-N
p-methoxystilbene,4-methoxystilbene,anisole, p-styryl,trans-4-methoxystilbene,e-4-methoxystilbene,1-methoxy-4-e-2-phenylethenyl benzene,1-p-methoxyphenyl-2-phenylethene,unii-5el148b7hg,benzene, 1-methoxy-4-2-phenylethenyl,1-methoxy-4-e-styryl-benzene
1245920
CHEBI:35094
1-methoxy-4-[(E)-2-phenylethenyl]benzene
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
1142-15-0
C15H14O
210.276
MFCD00017178
XWYXLYCDZKRCAD-BQYQJAHWSA-N
p-methoxystilbene,4-methoxystilbene,anisole, p-styryl,trans-4-methoxystilbene,e-4-methoxystilbene,1-methoxy-4-e-2-phenylethenyl benzene,1-p-methoxyphenyl-2-phenylethene,unii-5el148b7hg,benzene, 1-methoxy-4-2-phenylethenyl,1-methoxy-4-e-styryl-benzene
1245920
CHEBI:35094
1-methoxy-4-[(E)-2-phenylethenyl]benzene
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
Applications
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.

Solubility
Sparingly soluble (0.061 g/L) (25°C) in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 136°C to 138°C
Quantity 25g
Solubility Information Sparingly soluble (0.061g/L) (25°C) in water.
Formula Weight 210.27
Percent Purity 98%
Chemical Name or Material 4-Methoxystilbene

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 214-530-7

TSCA : No

Recommended Storage : Ambient temperatures