Alfa Aesar™ 4-Methoxytrityl chloride, 97%

Manufacturer: Alfa Aesar™ A1054530

Quantity 250g

 View more versions of this product

Catalog No. AAA1054530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14470-28-1
    Molecular Formula C20H17ClO
    Formula Weight 308.81
    MDL Number MFCD00000814
    Beilstein 798425
    Synonym p-Anisylchlorodiphenylmethane; 4-Methoxytriphenylmethyl chloride
    Chemical Name or Material 4-Methoxytrityl chloride

    Specifications

    Formula Weight 308.81
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 798425
    Melting Point 121° to 125°C
    Health Hazard 1 Warning
    Molecular Formula C20H17ClO
    MDL Number MFCD00000814
    CAS 14470-28-1
    Synonym p-Anisylchlorodiphenylmethane; 4-Methoxytriphenylmethyl chloride
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 4-Methoxytrityl chloride
    Assay 97%

    Description

    Applications
    It is used as selective protecting reagent that is used for primary hydroxyl groups.

    Solubility
    Soluble in toluene.

    Notes
    It is hygroscopic. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit