Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1332627-30-1
C11H13NO2
191.23
QOQHRSQQPZYCGY-UHFFFAOYSA-N
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
56766060
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
CC1CCNC2=C1C=C(C=C2)C(=O)O
1332627-30-1
C11H13NO2
191.23
QOQHRSQQPZYCGY-UHFFFAOYSA-N
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
56766060
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
CC1CCNC2=C1C=C(C=C2)C(=O)O
Melting Point 158°C to 160°C
Quantity 5g
Assay Percent Range 94% min. (HPLC)
Formula Weight 191.23
Percent Purity 95%
Chemical Name or Material 4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning