Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 158°C to 160°C |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 191.23 |
Percent Purity | 95% |
Chemical Name or Material | 4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction