Alfa Aesar™ 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers, 98%

Manufacturer: Alfa Aesar™ B2332822

Quantity 100g

 View more versions of this product

Catalog No. AAB2332822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19438-60-9
    Molecular Formula C9H12O3
    Formula Weight 168.19
    MDL Number MFCD00005927
    Synonym Hexahydro-4-methylphthalic anhydride
    Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers

    Specifications

    Formula Weight 168.19
    TSCA TSCA
    Molecular Formula C9H12O3
    MDL Number MFCD00005927
    Synonym Hexahydro-4-methylphthalic anhydride
    Health Hazard 1 Danger
    Boiling Point 119° to 121°C (1mmHg)
    Quantity 100g
    Flash Point >110°C (230°F)
    Odor Odorless
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P310-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 19438-60-9
    Health Hazard 2 GHS H Statement
    H334-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers
    Assay 98%

    GHS H Statement
    H334-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P310-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit