Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-1-naphthoyl chloride, 98%

Click to view available options
Quantity:
1g
5g
25g
87700-67-2
C12H9ClO
204.653
MFCD12025362
IXGMHYHWSPDNNS-UHFFFAOYSA-N
1-naphthalenecarbonylchloride, 4-methyl,1-naphthalenecarbonyl chloride, 4-methyl,4-methyl-1-naphthalenecarbonyl chloride,1-naphthalenecarbonyl chloride, 4-methyl-9ci,4-methyl-1-napthoyl chloride,4-methyl-1-naphthoyl chloride,4-methyl-naphthalene-1-carbonyl chloride
24887412
4-methylnaphthalene-1-carbonyl chloride
CC1=CC=C(C2=CC=CC=C12)C(=O)Cl
87700-67-2
C12H9ClO
204.653
MFCD12025362
IXGMHYHWSPDNNS-UHFFFAOYSA-N
1-naphthalenecarbonylchloride, 4-methyl,1-naphthalenecarbonyl chloride, 4-methyl,4-methyl-1-naphthalenecarbonyl chloride,1-naphthalenecarbonyl chloride, 4-methyl-9ci,4-methyl-1-napthoyl chloride,4-methyl-1-naphthoyl chloride,4-methyl-naphthalene-1-carbonyl chloride
24887412
4-methylnaphthalene-1-carbonyl chloride
CC1=CC=C(C2=CC=CC=C12)C(=O)Cl
Color Yellow
Quantity 25g
UN Number UN3261
Beilstein 3247818
Formula Weight 204.65
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Methyl-1-naphthoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.