Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-1-tetralone, 97%

Click to view available options
Quantity:
1g
5g
25g
19832-98-5
C11H12O
160.216
MFCD00001691
SRLHDEROUKFEMJ-UHFFFAOYSA-N
4-methyl-1-tetralone,4-methyl-3,4-dihydronaphthalen-1 2h-one,4-methyl-.alpha.-tetralone,1 2h-naphthalenone, 3,4-dihydro-4-methyl,4-methyl-1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-4-methyl-1 2h-naphthalenone,4-methyl-2,3,4-trihydronaphthalen-1-one,zlchem 108,pubchem11038,4-methyltetralin-1-one
89232
4-methyl-3,4-dihydro-2H-naphthalen-1-one
CC1CCC(=O)C2=CC=CC=C12
19832-98-5
C11H12O
160.216
MFCD00001691
SRLHDEROUKFEMJ-UHFFFAOYSA-N
4-methyl-1-tetralone,4-methyl-3,4-dihydronaphthalen-1 2h-one,4-methyl-.alpha.-tetralone,1 2h-naphthalenone, 3,4-dihydro-4-methyl,4-methyl-1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-4-methyl-1 2h-naphthalenone,4-methyl-2,3,4-trihydronaphthalen-1-one,zlchem 108,pubchem11038,4-methyltetralin-1-one
89232
4-methyl-3,4-dihydro-2H-naphthalen-1-one
CC1CCC(=O)C2=CC=CC=C12
Applications
4-Methyl-1-tetralone is an important intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 95°C (1mmHg)
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-tetralone