Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2,1,3-benzoxadiazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
29091-40-5
C7H6N2O
134.14
PTXJEYIAUIOTSH-UHFFFAOYSA-N
4-methylbenzo c 1,2,5 oxadiazole,4-methylbenzofurazan,4-methyl-benzofurazan,4-methyl-benzo 1,2,5 oxadiazole,2,1,3-benzoxadiazole, 4-methyl,methylbenzfurazan,2,1,3-benzoxadiazole,4-methyl,#
584114
4-methyl-2,1,3-benzoxadiazole
CC1=CC=CC2=NON=C12
29091-40-5
C7H6N2O
134.14
PTXJEYIAUIOTSH-UHFFFAOYSA-N
4-methylbenzo c 1,2,5 oxadiazole,4-methylbenzofurazan,4-methyl-benzofurazan,4-methyl-benzo 1,2,5 oxadiazole,2,1,3-benzoxadiazole, 4-methyl,methylbenzfurazan,2,1,3-benzoxadiazole,4-methyl,#
584114
4-methyl-2,1,3-benzoxadiazole
CC1=CC=CC2=NON=C12
Color Yellow
Quantity 5g
Melting Point 40.0°C to 41.0°C
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 134.14
Physical Form Crystalline Powder or Lms
Percent Purity 95%
Chemical Name or Material 4-Methyl-2, 1, 3-benzoxadiazole
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning