Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methyl-2,1,3-benzoxadiazole, 95%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Specifications
Packaging | Glass Bottle |
Melting Point | 40.0°C to 41.0°C |
Quantity | 1g |
Infrared Spectrum | Authentic |
Color | Yellow |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Assay Percent Range | 96% min. (GC) |
Formula Weight | 134.14 |
Physical Form | Crystalline Powder or Lms |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning