Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-2-(2-thiazolylazo)phenol, 97%

Manufacturer:  Alfa Aesar™ H5554414

Quantity 25g

 View more versions of this product

Catalog No. AAH5554414

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1823-44-5
MFCD00005323
2-(2-Thiazolylazo)-p-cresol
219.26
190372
4-Methyl-2-(2-thiazolylazo)phenol

Specifications

Orange to brown
25g
Solid
MFCD00005323
2-(2-Thiazolylazo)-p-cresol
97%
130° to 132°C
1823-44-5
219.26
190372
4-Methyl-2-(2-thiazolylazo)phenol

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit