Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-nitrobenzonitrile, 99%, Acros Organics

Click to view available options
Quantity:
5g
26830-95-5
C8H6N2O2
162.148
MFCD00056107
QGBSLPHQCUIZKK-UHFFFAOYSA-N
2-nitro-p-tolunitrile,2-nitro-4-toluonitrile,benzonitrile, 4-methyl-2-nitro,2-nitro-4-methylbenzonitrile,4-methyl-2-nitro-benzonitrile,4-methyl-2-nitrobenzenecarbonitrile,pubchem13730,acmc-1cjm6,ksc494k5h,4-methyl-2-nitro benzonitrile
96235
4-methyl-2-nitrobenzonitrile
CC1=CC(=C(C=C1)C#N)[N+](=O)[O-]
26830-95-5
C8H6N2O2
162.148
MFCD00056107
QGBSLPHQCUIZKK-UHFFFAOYSA-N
2-nitro-p-tolunitrile,2-nitro-4-toluonitrile,benzonitrile, 4-methyl-2-nitro,2-nitro-4-methylbenzonitrile,4-methyl-2-nitro-benzonitrile,4-methyl-2-nitrobenzenecarbonitrile,pubchem13730,acmc-1cjm6,ksc494k5h,4-methyl-2-nitro benzonitrile
96235
4-methyl-2-nitrobenzonitrile
CC1=CC(=C(C=C1)C#N)[N+](=O)[O-]
Infrared Spectrum Authentic
Color Brown
Quantity 5g
Melting Point 96°C to 100°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula O2NC6H3(CH3)CN
Beilstein 09, 501
Formula Weight 162.15
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Methyl-2-nitrobenzonitrile, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning