Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-2-nitrophenyl isothiocyanate, 95%

Click to view available options
Quantity:
1g
5g
17614-74-3
C8H6N2O2S
194.208
MFCD00041248
CUQUTWSUXWDJKF-UHFFFAOYSA-N
4-methyl-2-nitrophenyl isothiocyanate, 4-methyl-2-nitrophenylisothiocyanate, benzene,1-isothiocyanato-4-methyl-2-nitro, 4-methyl-2-nitrobenzenisothiocyanate, acmc-1brfl, 4-methy-2-nitrophenyl isothiocyanate, 1-isothiocyanato-4-methyl-2-nitro-benzene, #
140258
1-isothiocyanato-4-methyl-2-nitrobenzene
CC1=CC(=C(C=C1)N=C=S)[N+](=O)[O-]
17614-74-3
C8H6N2O2S
194.208
MFCD00041248
CUQUTWSUXWDJKF-UHFFFAOYSA-N
4-methyl-2-nitrophenyl isothiocyanate, 4-methyl-2-nitrophenylisothiocyanate, benzene,1-isothiocyanato-4-methyl-2-nitro, 4-methyl-2-nitrobenzenisothiocyanate, acmc-1brfl, 4-methy-2-nitrophenyl isothiocyanate, 1-isothiocyanato-4-methyl-2-nitro-benzene, #
140258
1-isothiocyanato-4-methyl-2-nitrobenzene
CC1=CC(=C(C=C1)N=C=S)[N+](=O)[O-]
Sensitivity Moisture sensitive
Quantity 1g
Melting Point 62°C to 67°C
Odor Irritating
UN Number UN2923
Beilstein 2969500
Formula Weight 194.21
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-nitrophenyl isothiocyanate

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H331-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures