Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
332360-07-3,4502-00-5
C6H9NaO3·xH2O
152.13
MFCD00150776
FUDIEMDWYRETTC-UHFFFAOYSA-M
4-methyl-2-oxopentanoic acid sodium salt hydrate,sodium hydrate ?-ketoisocaproate,4-methyl-2-oxopentanoic acid,sodium salt,hydrate,98+%
23680490
sodium;4-methyl-2-oxopentanoate;hydrate
CC(C)CC(=O)C(=O)[O-].O.[Na+]
332360-07-3,4502-00-5
C6H9NaO3·xH2O
152.13
MFCD00150776
FUDIEMDWYRETTC-UHFFFAOYSA-M
4-methyl-2-oxopentanoic acid sodium salt hydrate,sodium hydrate ?-ketoisocaproate,4-methyl-2-oxopentanoic acid,sodium salt,hydrate,98+%
23680490
sodium;4-methyl-2-oxopentanoate;hydrate
CC(C)CC(=O)C(=O)[O-].O.[Na+]
Quantity 1g
Packaging Glass bottle
Melting Point 280°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3)2CHCH2COCO2Na·xH2O
Solubility Information Solubility in water: soluble
Formula Weight 152.13
Percent Purity 98+%
Chemical Name or Material 4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%
Show More Show Less