Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentanol, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
108-11-2
C6H14O
102.18
MFCD00004550
WVYWICLMDOOCFB-UHFFFAOYSA-N
4-methyl-2-pentanol,2-pentanol, 4-methyl,isobutylmethylcarbinol,2-methyl-4-pentanol,methyl amyl alcohol,methyl isobutyl carbinol,mibc,1,3-dimethylbutanol,4-methylpentanol-2,isobutylmethylmethanol
7910
4-methylpentan-2-ol
CC(C)CC(C)O
108-11-2
C6H14O
102.18
MFCD00004550
WVYWICLMDOOCFB-UHFFFAOYSA-N
4-methyl-2-pentanol,2-pentanol, 4-methyl,isobutylmethylcarbinol,2-methyl-4-pentanol,methyl amyl alcohol,methyl isobutyl carbinol,mibc,1,3-dimethylbutanol,4-methylpentanol-2,isobutylmethylmethanol
7910
4-methylpentan-2-ol
CC(C)CC(C)O
Infrared Spectrum Authentic
Specific Gravity 0.8
Color Colorless
Density 0.8000g/mL
Quantity 10L
Flash Point 41°C
Melting Point -90.0°C
Refractive Index 1.4100 to 1.4120
Boiling Point 132.0°C
Packaging Plastic drum
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (CH3)2CHCH2CH(OH)CH3
Beilstein 01, 410
Solubility Information Solubility in water: 20g/L. Other solubilities: misscible with most common organic solvents
Formula Weight 102.18
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material 4-Methyl-2-pentanol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning