Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-2-pentanone, ACS, 98.5+%

Manufacturer:  Alfa Aesar™ 33346AY

Quantity 4 x 500mL

 View more versions of this product

Catalog No. AA33346AY

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

108-10-1
MFCD00008938
605399
Methyl isobutyl ketone; Isopropyl acetone
ACS Grade
100.16
UN1245
14,5207
4-Methyl-2-pentanone

Specifications

117° to 118°C
108-10-1
Liquid
MFCD00008938
605399
14,5207
Colorless
13° (55°F)
4-Methyl-2-pentanone
ACS Grade
4 x 500mL
1.3960
100.16
UN1245
0.801g/mL
Methyl isobutyl ketone; Isopropyl acetone
−84°C
Mild
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ISOBUTYL KETONE

RTECSNumber : SA9275000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit