Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentyne, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
21020-27-9
C6H10
82.14
MFCD00041615
SLMFWJQZLPEDDU-UHFFFAOYSA-N
4-methyl-2-pentyne,2-pentyne, 4-methyl,isopropylmethylacetylene,acmc-1co71
140789
4-methylpent-2-yne
CC#CC(C)C
21020-27-9
C6H10
82.14
MFCD00041615
SLMFWJQZLPEDDU-UHFFFAOYSA-N
4-methyl-2-pentyne,2-pentyne, 4-methyl,isopropylmethylacetylene,acmc-1co71
140789
4-methylpent-2-yne
CC#CC(C)C
Melting Point -110°C
Specific Gravity 0.716
Boiling Point 71°C to 73°C
Flash Point -5°C
Density 0.7160g/mL
Quantity 1g
Color Yellow
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080 (20°C, 589nm)
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3C∽CCH(CH3)2
Formula Weight 82.14
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 4-Methyl-2-pentyne
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger