Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-2-phenylthiazole, 98%

Click to view available options
Quantity:
1g
5g
1826-17-1
C10H9NS
175.249
MFCD00272328
IPOHWQDCODUHTD-UHFFFAOYSA-N
4-methyl-2-phenylthiazole,thiazole, 4-methyl-2-phenyl,acmc-1brhd,4-methyl-2-phenyl-thiazole,thiazole,4-methyl-2-phenyl,#
551876
4-methyl-2-phenyl-1,3-thiazole
CC1=CSC(=N1)C2=CC=CC=C2
1826-17-1
C10H9NS
175.249
MFCD00272328
IPOHWQDCODUHTD-UHFFFAOYSA-N
4-methyl-2-phenylthiazole,thiazole, 4-methyl-2-phenyl,acmc-1brhd,4-methyl-2-phenyl-thiazole,thiazole,4-methyl-2-phenyl,#
551876
4-methyl-2-phenyl-1,3-thiazole
CC1=CSC(=N1)C2=CC=CC=C2
Refractive Index 1.627
Quantity 1g
Formula Weight 175.25
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-phenylthiazole

TSCA : No

Recommended Storage : Ambient temperatures