Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiophene-2-boronic acid, 96%, Acros Organics™

Click to view available options
Quantity:
500mg
5g
Packaging:
Glass bottle
162607-15-0
C5H7BO2S
141.979
DFUMIZDUIJNUJU-UHFFFAOYSA-N
4-methylthiophene-2-boronic acid,4-methylthiophen-2-yl boronic acid,4-methyl-2-thienylboric acid,4-methyl-2-thienylboronic acid,4-methylthiophene 2-boronic acid,4-methyl-2-thienyl boronic acid,4-methylthiophen-2-yl boranediol,boronic acid, 4-methyl-2-thienyl,4-methylthiophene-2-boronicacid,acmc-209dok
2734373
(4-methylthiophen-2-yl)boronic acid
B(C1=CC(=CS1)C)(O)O
162607-15-0
C5H7BO2S
141.979
DFUMIZDUIJNUJU-UHFFFAOYSA-N
4-methylthiophene-2-boronic acid,4-methylthiophen-2-yl boronic acid,4-methyl-2-thienylboric acid,4-methyl-2-thienylboronic acid,4-methylthiophene 2-boronic acid,4-methyl-2-thienyl boronic acid,4-methylthiophen-2-yl boranediol,boronic acid, 4-methyl-2-thienyl,4-methylthiophene-2-boronicacid,acmc-209dok
2734373
(4-methylthiophen-2-yl)boronic acid
B(C1=CC(=CS1)C)(O)O
Color White to Beige
Packaging Glass bottle
Quantity 500mg
Melting Point 125.0°C to 126.0°C
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 141.99
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 4-Methylthiophene-2-boronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.