Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
119-32-4
C7H8N2O2
152.15
MFCD00007910
GDIIPKWHAQGCJF-UHFFFAOYSA-N
4-amino-2-nitrotoluene,3-nitro-4-methylaniline,3-nitro-p-toluidine,m-nitro-p-toluidine,2-nitro-4-aminotoluene,benzenamine, 4-methyl-3-nitro,5-nitro-4-toluidine,p-toluidine, 3-nitro,gl-amin czech,gl-amin
8390
CHEBI:81670
4-methyl-3-nitroaniline
CC1=C(C=C(C=C1)N)[N+](=O)[O-]
119-32-4
C7H8N2O2
152.15
MFCD00007910
GDIIPKWHAQGCJF-UHFFFAOYSA-N
4-amino-2-nitrotoluene,3-nitro-4-methylaniline,3-nitro-p-toluidine,m-nitro-p-toluidine,2-nitro-4-aminotoluene,benzenamine, 4-methyl-3-nitro,5-nitro-4-toluidine,p-toluidine, 3-nitro,gl-amin czech,gl-amin
8390
CHEBI:81670
4-methyl-3-nitroaniline
CC1=C(C=C(C=C1)N)[N+](=O)[O-]
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 74°C to 78°C
Color Brown or Ochre to Orange
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H3(NO2)NH2
Beilstein 12, 996
Formula Weight 152.15
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Methyl-3-nitroaniline
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning