Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-3-nitrophenol, 98%

Manufacturer:  Alfa Aesar™ 4428504

Quantity 2g

 View more versions of this product

Catalog No. AA4428504


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2042-14-0
153.14
UN2446
4-Methyl-3-nitrophenol
C7H7NO3
MFCD00007244
3-nitro-p-cresol

Specifications

2042-14-0
C7H7NO3
MFCD00007244
3-nitro-p-cresol
2g
4-Methyl-3-nitrophenol
Powder
153.14
UN2446
77° to 81°C
Phenol-like
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit