Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)benzonitrile, 97+%

Click to view available options
Quantity:
1g
5g
261952-06-1
C9H6F3N
185.149
MFCD01631597
YWAGJCPTOFPGIS-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzonitrile,3-trifluoromethyl-4-methylbenzonitrile,3-trifluoromethyl-4-methyl benzonitrile,3-trifluoromethyl-p-tolunitrile,5-cyano-2-methylbenzotrifluoride,benzonitrile, 4-methyl-3-trifluoromethyl,4-methyl-3-trifluoromethyl benzenecarbonitrile,pubchem21353,acmc-20agl9,2-methyl-5-cyanobenzotrifluoride
2775598
4-methyl-3-(trifluoromethyl)benzonitrile
CC1=C(C=C(C=C1)C#N)C(F)(F)F
261952-06-1
C9H6F3N
185.149
MFCD01631597
YWAGJCPTOFPGIS-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzonitrile,3-trifluoromethyl-4-methylbenzonitrile,3-trifluoromethyl-4-methyl benzonitrile,3-trifluoromethyl-p-tolunitrile,5-cyano-2-methylbenzotrifluoride,benzonitrile, 4-methyl-3-trifluoromethyl,4-methyl-3-trifluoromethyl benzenecarbonitrile,pubchem21353,acmc-20agl9,2-methyl-5-cyanobenzotrifluoride
2775598
4-methyl-3-(trifluoromethyl)benzonitrile
CC1=C(C=C(C=C1)C#N)C(F)(F)F
Quantity 1g
Melting Point 60°C to 62°C
UN Number UN3439
Formula Weight 185.15
Percent Purity ≥97%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.