Alfa Aesar™ 4-Methyl-3-(trifluoromethyl)phenol, 97%

Manufacturer: Alfa Aesar™ H32051MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32051MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H7F3O
    Formula Weight 176.14
    MDL Number MFCD09832294
    UN Number UN3261
    Synonym 5-Hydroxy-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-cresol
    Chemical Name or Material 4-Methyl-3-(trifluoromethyl)phenol

    Specifications

    Formula Weight 176.14
    Molecular Formula C8H7F3O
    MDL Number MFCD09832294
    UN Number UN3261
    Synonym 5-Hydroxy-2-methylbenzotrifluoride; 3-Trifluoromethyl-p-cresol
    Health Hazard 1 Danger
    Melting Point 45° to 48°C
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4-Methyl-3-(trifluoromethyl)phenol
    Assay 97%

    Description

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H314-H318-H302-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit