Alfa Aesar™ 4-Methyl-4-methylthio-2-pentanone, 99%

Manufacturer: Alfa Aesar™ B2205814

Quantity 25g

 View more versions of this product

Catalog No. AAB2205814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23550-40-5
    Molecular Formula C6H8OS
    Formula Weight 146.25
    MDL Number MFCD00049253
    Synonym 4-Methyl-4-(methylmercapto)pentan-2-one
    Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone

    Specifications

    Formula Weight 146.25
    Health Hazard 1 Warning
    Boiling Point 80° to 82°C (10mmHg)
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C6H8OS
    MDL Number MFCD00049253
    CAS 23550-40-5
    Synonym 4-Methyl-4-(methylmercapto)pentan-2-one
    Flash Point 72°C (162°F)
    Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone
    Assay 99%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit