Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-4-methylthio-2-pentanone, 99%

Click to view available options
Quantity:
25g
100g
23550-40-5
C7H14OS
146.248
MFCD00049253
DHANVOSHQILVNQ-UHFFFAOYSA-N
4-methylthio-4-methyl-2-pentanone,4-methyl-4-methylthio pentan-2-one,2-pentanone, 4-methyl-4-methylthio,4-methyl-4-methylthio-2-pentanone,unii-g69hk72q3y,4-methyl-4-methylsulfanyl pentan-2-one,cassis pentanone,fema no. 3376,corps cassis
4682590
4-methyl-4-methylsulfanylpentan-2-one
CC(=O)CC(C)(C)SC
23550-40-5
C7H14OS
146.248
MFCD00049253
DHANVOSHQILVNQ-UHFFFAOYSA-N
4-methylthio-4-methyl-2-pentanone,4-methyl-4-methylthio pentan-2-one,2-pentanone, 4-methyl-4-methylthio,4-methyl-4-methylthio-2-pentanone,unii-g69hk72q3y,4-methyl-4-methylsulfanyl pentan-2-one,cassis pentanone,fema no. 3376,corps cassis
4682590
4-methyl-4-methylsulfanylpentan-2-one
CC(=O)CC(C)(C)SC
Quantity 100g
Boiling Point 80°C to 82°C (10mmHg)
Flash Point 72°C (162°F)
Formula Weight 146.25
Percent Purity 99%
Chemical Name or Material 4-Methyl-4-methylthio-2-pentanone

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA