Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyl-5-nonanol, 98%

Click to view available options
Quantity:
10g
50g
250g
154170-44-2
C10H22O
158.285
MFCD00046696
MBZNNOPVFZCHID-UHFFFAOYSA-N
4-methyl-5-nonanol,5-nonanol, 4-methyl,4-methyl 5-nonanol,acmc-1c1h3
3025865
4-methylnonan-5-ol
CCCCC(C(C)CCC)O
154170-44-2
C10H22O
158.285
MFCD00046696
MBZNNOPVFZCHID-UHFFFAOYSA-N
4-methyl-5-nonanol,5-nonanol, 4-methyl,4-methyl 5-nonanol,acmc-1c1h3
3025865
4-methylnonan-5-ol
CCCCC(C(C)CCC)O
Quantity 50g
Formula Weight 158.29
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-nonanol