Alfa Aesar™ 4-Methyl-5-nonanone, 99+%

Manufacturer: Alfa Aesar™ B2321922

Quantity 100g

 View more versions of this product

Catalog No. AAB2321922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35900-26-6
    Molecular Formula C10H20O
    Formula Weight 156.27
    MDL Number MFCD00048807
    Chemical Name or Material 4-Methyl-5-nonanone

    Specifications

    Formula Weight 156.27
    MDL Number MFCD00048807
    TSCA TSCA
    Molecular Formula C10H20O
    CAS 35900-26-6
    Boiling Point 189° to 190°C
    Quantity 100g
    Chemical Name or Material 4-Methyl-5-nonanone
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit